CAS RN 7367-88-6
IUPAC Name:
ethyl (2E)-dec-2-enoate
Synonyms:
ethyl (2E)-dec-2-enoate
Molecular Weight (g/mol):
198.31
Molecular Formula:
C12H22O2
InChi Key:
GNJARWZWODMTDR-ZHACJKMWSA-N
SMILES:
CCCCCCC\C=C\C(=O)OCC
1
–
1
of
1
results
1
Ethyl trans-2-decenoate, 97%, Thermo Scientific Chemicals
CAS: 7367-88-6 Molecular Formula: C12H22O2 Molecular Weight (g/mol): 198.306 MDL Number: MFCD00015299 InChI Key: GNJARWZWODMTDR-ZHACJKMWSA-N Synonym: ethyl trans-2-decenoate,ethyl 2-decenoate,ethyl e-2-decenoate,trans-2-decenoic acid ethyl ester,ethyl 2e-dec-2-enoate,2-decenoic acid, ethyl ester, 2e,unii-b2k05s6l5q,2-decenoic acid, ethyl ester, e,ethyl 2-decenoate, 2e,fema no. 3641 PubChem CID: 5463904 ChEBI: CHEBI:87324 IUPAC Name: ethyl (E)-dec-2-enoate SMILES: CCCCCCCC=CC(=O)OCC