accessibility menu, dialog, popup

UserName

CAS RN 756520-57-7

BOOH3CCH3H3CH3CNNH2O

CAS RN 756520-57-7

IUPAC Name: 3-(benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: 3-(benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 326.2
Molecular Formula: C18H23BN2O3
InChi Key: RPQADWBEACHUFT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(OCC2=CC=CC=C2)=C(N)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results