accessibility menu, dialog, popup

UserName

NNH2NClHCl

CAS RN 77145-35-8

IUPAC Name: 1-(3-chloropyridin-2-yl)piperidin-4-amine hydrochloride
Synonyms: 1-(3-chloropyridin-2-yl)piperidin-4-amine hydrochloride
Molecular Weight (g/mol): 248.15
Molecular Formula: C10H15Cl2N3
InChi Key: NOSKSNWYGYGGBH-UHFFFAOYSA-N
SMILES: Cl.NC1CCN(CC1)C1=C(Cl)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results