accessibility menu, dialog, popup

UserName

NaOAsO

CAS RN 7784-46-5

IUPAC Name: sodium oxoarsinite
Synonyms: sodium arsenite
Molecular Weight (g/mol): 129.91
Molecular Formula: AsNaO2
InChi Key: PTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES: [Na+].[O-][As]=O
Molecular Weight (g/mol) 
  • (8)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (4)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (8)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)