accessibility menu, dialog, popup

UserName

CAS RN 85-32-5

NH2NONNOOPHOOHOOHOHHN

CAS RN 85-32-5

IUPAC Name: {[5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: guanosine monophosphate
Molecular Weight (g/mol): 363.22
Molecular Formula: C10H14N5O8P
InChi Key: RQFCJASXJCIDSX-UHFFFAOYNA-N
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(COP(O)(O)=O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results