accessibility menu, dialog, popup

UserName

CAS RN 850568-67-1

BCH3H3CNHOOH

CAS RN 850568-67-1

IUPAC Name: [4-(1-cyano-1-methylethyl)phenyl]boronic acid
Synonyms: 4-(1-cyano-1-methylethyl)phenylboronic acid
Molecular Weight (g/mol): 189.02
Molecular Formula: C10H12BNO2
InChi Key: BNXBFDTWYHAEIR-UHFFFAOYSA-N
SMILES: CC(C)(C#N)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results