accessibility menu, dialog, popup

UserName

H3CSONHNNFHCl

CAS RN 869185-85-3

IUPAC Name: 4-[4-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-1H-imidazol-5-yl]pyridine hydrochloride
Synonyms: 4-[5-(4-fluorophenyl)-2-(4-methanesulfinylphenyl)-3H-imidazol-4-yl]pyridine hydrochloride
Molecular Weight (g/mol): 413.9
Molecular Formula: C21H17ClFN3OS
InChi Key: WOSGGXINSLMASH-UHFFFAOYNA-N
SMILES: Cl.CS(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=NC=C1)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (4)