CAS RN 876707-99-2
CAS RN 876707-99-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
MedChemExpress Nucleoside-Analog-1
MedChemExpress Nucleoside-Analog-1 is a 4′-Azidocytidine analogue against Hepatitis C virus replication.
2
MedChemExpress Nucleoside-Analog-1
MedChemExpress Nucleoside-Analog-1 is a 4′-Azidocytidine analogue against Hepatitis C virus replication.
3
MedChemExpress Nucleoside-Analog-1
MedChemExpress Nucleoside-Analog-1 is a 4′-Azidocytidine analogue against Hepatitis C virus replication.
4
MedChemExpress Nucleoside-Analog-1
MedChemExpress Nucleoside-Analog-1 is a 4′-Azidocytidine analogue against Hepatitis C virus replication.