CAS RN 87771-41-3
CAS RN 87771-41-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(3-(2-Hydroxypropan-2-yl)phenyl)ethanone, TRC
CAS: 87771-41-3 Molecular Formula: C11H14O2 Molecular Weight (g/mol): 178.23 Synonym: 1-[3-(1-Hydroxy-1-methylethyl)phenyl]ethanone,1-(3-(2-Hydroxypropan-2-yl)phenyl)ethanone,1-[3-(2-Hydroxypropan-2-yl)phenyl]ethan-1-one; IUPAC Name: 1-[3-(2-hydroxypropan-2-yl)phenyl]ethanone SMILES: CC(=O)c1cccc(c1)C(C)(C)O