missing translation for 'onlineSavingsMsg'
Learn More

1-(3-(2-Hydroxypropan-2-yl)phenyl)ethanone, TRC

Product Code. 30007802
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007802 100 mg 100mg
30004580 500 mg 500mg
30007255 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007802 Supplier Toronto Research Chemicals Supplier No. TRCH853270100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87771-41-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
Synonym 1-[3-(1-Hydroxy-1-methylethyl)phenyl]ethanone, 1-(3-(2-Hydroxypropan-2-yl)phenyl)ethanone, 1-[3-(2-Hydroxypropan-2-yl)phenyl]ethan-1-one;
IUPAC Name 1-[3-(2-hydroxypropan-2-yl)phenyl]ethanone
SMILES CC(=O)c1cccc(c1)C(C)(C)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H14O2/c1-8(12)9-5-4-6-10(7-9)11(2,3)13/h4-7,13H,1-3H3
Formula Weight 178.0994
Chemical Name or Material 1-(3-(2-Hydroxypropan-2-yl)phenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.