CAS RN 98488-10-9
IUPAC Name:
2,5-dimethyl-1H,7H-pyrazolo[1,5-a]pyrimidin-7-one
Synonyms:
2,5-dimethyl-1H-pyrazolo[1,5-a]pyrimidin-7-one
Molecular Weight (g/mol):
163.18
Molecular Formula:
C8H9N3O
InChi Key:
LGQMDCIHBYTQJU-UHFFFAOYSA-N
SMILES:
CC1=CC2=NC(C)=CC(=O)N2N1
1
–
1
of
1
results
1
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7(4H)-one, 97%, Thermo Scientific™
CAS: 98488-10-9 Molecular Formula: C8H9N3O Molecular Weight (g/mol): 163.18 MDL Number: MFCD08729601 InChI Key: LGQMDCIHBYTQJU-UHFFFAOYSA-N Synonym: 2,5-dimethylpyrazolo 1,5-a pyrimidin-7-ol,2,5-dimethylpyrazolo 1,5-a pyrimidin-7 4h-one,2,5-dimethyl-4h-pyrazolo 1,5-a pyrimidin-7-one,2,5-dimethyl-4,7-dihydropyrazolo 1,5-a pyrimidin-7-one,2,5-dimethyl-1h-pyrazolo 1,5-a pyrimidin-7-one,2,5-dimethylpyrazolo 1,5-a pyrimidin-7-one,2,5-dimethyl-4h,7h-pyrazolo 1,5-a pyrimidin-7-one,acmc-20aoa7,maybridge1_004141 PubChem CID: 736199 IUPAC Name: 2,5-dimethyl-1H-pyrazolo[1,5-a]pyrimidin-7-one SMILES: CC1=CC2=NC(C)=CC(=O)N2N1