missing translation for 'onlineSavingsMsg'
Learn More

1-Oxo Ibuprofen, TRC

Product Code. 30005571
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005571 100 mg 100mg
30004283 10 mg 10mg
30010961 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005571 Supplier Toronto Research Chemicals Supplier No. TRCO870035100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65813-55-0
Molecular Formula C13 H16 O3
Molecular Weight (g/mol) 220.26
Synonym (2RS)-2-[4-(2-Methyl-propanoyl)phenyl]propanoic Acid, Ibuprofen Imp. J (EP)
IUPAC Name 2-[4-(2-methylpropanoyl)phenyl]propanoic acid
SMILES CC(C)C(=O)c1ccc(cc1)C(C)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H16O3/c1-8(2)12(14)11-6-4-10(5-7-11)9(3)13(15)16/h4-9H,1-3H3,(H,15,16)
Formula Weight 220.1099
Percent Purity >95%
Chemical Name or Material (2RS)-2-[4-(2-Methyl-propanoyl)phenyl]propanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.