Learn More
Fisherbrand™ 13mm PP Screw Seal, White, Center hole, 13-425 thread, Assembled septum
Product Code. 14861641 Shop All Fisherbrand Products
Click to view available options
Color:
Blue, White
Unit Size:
Pack of 1000
Product Code. 14861641
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Polypropylene caps are chemically inert and suitable for most chromatography applications.
  • 13mm
  • PP
  • Screw Cap
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) Silicone/PTFE
Color Blue, White
Diameter (Metric) Outer 13 mm
Material Polypropylene
Pre-slit Yes
For Use With (Application) HPLC
Quantity 100
Thickness (Metric) 1.5 mm
Product Type Screw Cap