missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,1,1-trimethoxyethane, TRC

Product Code. 30354967
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354967 5 g 5g
30349516 25 g 25g
30360661 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354967 Supplier Toronto Research Chemicals Supplier No. TRCC4217805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74974-54-2
Molecular Formula C5H11ClO3
Molecular Weight (g/mol) 154.59
Synonym 1-Chloro-2,2,2-trimethoxyethane, Trimethyl (Chloromethyl)orthoformate, Trimethyl Orthochloroacetate
IUPAC Name 2-chloro-1,1,1-trimethoxyethane
SMILES ClCC(OC)(OC)OC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H11ClO3/c1-7-5(4-6,8-2)9-3/h4H2,1-3H3
Formula Weight 154.0397
Chemical Name or Material 2-Chloro-1,1,1-trimethoxyethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.