Learn More
Fisherbrand™ 20mm Crimp Seal, Green, Aluminum Cap, 10mm Center hole, Assembled septum
Shop All Fisherbrand Products
Click to view available options
Color:
Dark Gray, Green Lacquered
Unit Size:
Pack of 1000
Specifications
Specifications
| Material (Liner) | Butyl |
| Color | Dark Gray, Green Lacquered |
| Diameter (Metric) Outer | 20 mm |
| Material | Aluminum |
| For Use With (Application) | Headspace |
| Quantity | 100 |
| Thickness (Metric) | 3 mm |
| Type | Crimp Cap |