Learn More
4-Chlorobutyrophenone, 95%, Thermo Scientific™
Product Code. p-7044432
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11362496
Brand: Thermo Scientific Alfa Aesar L06383.06
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 939-52-6
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00001005
InChI Key GHEFQKHLHFXSBR-UHFFFAOYSA-N
Synonym 4-chlorobutyrophenone, gamma-chlorobutyrophenone, 4-chloro-1-oxo-1-phenylbutane, .gamma.-chlorobutyrophenone, 1-butanone,4-chloro-1-phenyl, chlorpropylphenylketon, 4-chlorobutyrylbenzene, 4-chlorobutyro-phenone, 3-benzoylpropyl chloride, .omega.-chlorobutyrophenone
PubChem CID 253533
IUPAC Name 4-chloro-1-phenylbutan-1-one
SMILES ClCCCC(=O)C1=CC=CC=C1
Specifications
Melting Point 19°C to 20°C
Density 1.137
Boiling Point 138°C to 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 5 g
Beilstein 878974
Formula Weight 182.65
Percent Purity 95%
Chemical Name or Material 4-Chlorobutyrophenone