missing translation for 'onlineSavingsMsg'
Learn More

Allyl(chloromethyl)dimethylsilane, 97+%, Thermo Scientific™

Product Code. 11364937
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364937 1 g 1g
11354937 5 g 5g
11374937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364937 Supplier Thermo Scientific Alfa Aesar Supplier No. 041953.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75422-66-1
Molecular Formula C6H13ClSi
Molecular Weight (g/mol) 148.71
MDL Number MFCD00012045
InChI Key UHHLOVCFFWGSMO-UHFFFAOYSA-N
Synonym allyl chloromethyl dimethylsilane, silane, chloromethyl dimethyl-2-propenyl, chloromethyl dimethylprop-2-en-1-ylsilane, acmc-1bfca, dimethylallylchloromethylsilane, 3-chloromethyldimethylsilyl-1-propene, allyl chloromethyl dimethyl dimethylsilane, silane,allyl, chloromethyl ,dimethyl, chloromethyl dimethyl prop-2-en-1-yl silane
PubChem CID 556526
IUPAC Name chloromethyl-dimethyl-prop-2-enylsilane
SMILES C[Si](C)(CCl)CC=C

Specifications

Boiling Point 165°C (50mmHg)
Physical Form Liquid
Quantity 1 g
UN Number UN1993
Flash Point 27°C (81°F)
Formula Weight 148.71
Percent Purity ≥97%
Refractive Index 1.449
Sensitivity Moisture sensitive
Density 0.907 g/mL
Chemical Name or Material Allyl(chloromethyl)dimethylsilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.