missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hexacyanoferrate(Ii) Hydrate, ≥92.0% anhydrous basis (RT), Solstice

Product Code. 15610910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Poly Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15610910 250 g Poly Bottle 250g
15620910 50 g Poly Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15610910 Supplier Honeywell-Fluka Supplier No. 09810250G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spot test reagent, for the determination of Ca,≥92.0% anhydrous basis (RT), ≤15% water

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14481-29-9
Molecular Formula C6H16FeN10
Molecular Weight (g/mol) 284.109
MDL Number MFCD00149957
InChI Key ZXQVPEBHZMCRMC-UHFFFAOYSA-R
Synonym ammonium ferrocyanide, ammonium hexacyanoferrate ii, ammoniumferrocyanid, tetraammonium ferrocyanide, ammonium hexacyanoferrate 4-, tetraammonium hexacyanidoferrate, ammonium hexacyanidoferrate ii, ammonium hexacyanidoferrate 4-, nh4 4 fe cn 6
PubChem CID 17756740
IUPAC Name tetraazanium;iron(2+);hexacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[NH4+].[NH4+].[NH4+].[Fe+2]

Specifications

Color Green to White
Quantity 250 g
Linear Formula (NH4)4[Fe(CN)6] · xH2O
UN Number NONH for all modes of transport
Formula Weight 284.10g/mol (anhydrous basis)
Percent Purity ≥92.0% anhydrous basis (RT)
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Ammonium Hexacyanoferrate(II) Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.