Learn More
Kimble Chase™ Borosilicate Glass Serum Vials without Closures
Kimble Chase™ Borosilicate Glass Serum Vials without Closures are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, vaccines, blood plasma, culture media, chromatography samples, and more.
Brand:  Kimble Chase™ 62121D-5
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10023444
  • € 536.00 / Pack of 864
Estimated Shipment: 26-06-2023
to see stock.
Add to basket
Description
Description
  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Autoclavable
  • Aluminum seals, stoppers and septa are available separately
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
23 mm
5 mL
47 mm
Borosilicate Glass (Type 1 Class A)
864
A/S 20
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers