Description
- Timer with full number keyboard
- 76 x 63 x 25mm
- 10 hours
Specifications
Specifications
| Product Type | Countdown Timer |
| Alarms | Audible |
| Display | Digital |
| Dimensions (L x W x H) | 60 W x 60 H mm |
| Width (Metric) | 64 mm |
| Height (Metric) | 60 mm |
| Quantity | 1 Each |