missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Cryo.s Round Bottom 2 mL Polypropylene Tubes with External Thread Cap
Suitable for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.
€ 272.00
Specifications
Diameter (Metric) | 12.4 mm |
---|---|
Product Type | Cryogenic Storage Tube |
Capacity (Metric) | 2 mL |
Bottom Shape | Round |
Material | Polypropylene |
Product Code | Brand | Closure Color | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Includes | Price | Quantity & Availability | ||||
10751761
|
Greiner Bio-One
126261 |
Natural | Starfoot Base, 150 white cap inserts |
€ 272.00
Pack of 500 |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- With Star foot base
- Without ID writing area
- Sterile
- Non-pyrogenic and non-toxic
- Includes 150 white cap inserts
- Polyethylener case
- Dia. x Height: 12.4 x 47mm
Specifications
12.4 mm | |
2 mL | |
Polypropylene | |
47 mm | |
Sterile |
Cryogenic Storage Tube | |
Round | |
External | |
Polyethylene case | |
Threaded |