Learn More
Ethoxytriethylsilane, 97%
Product Code. 11349305
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11349305
Brand: Thermo Scientific Alfa Aesar L16656.06
Request Bulk or Custom Format
CAS: 597-67-1 | C8H20OSi | 160.332 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 597-67-1
Molecular Formula C8H20OSi
Molecular Weight (g/mol) 160.332
MDL Number MFCD00054718
InChI Key DFJDZTPFNSXNAX-UHFFFAOYSA-N
Synonym triethylethoxysilane, silane, ethoxytriethyl, ethoxy triethyl silane, acmc-1al7t
PubChem CID 284135
IUPAC Name ethoxy(triethyl)silane
SMILES CCO[Si](CC)(CC)CC
Specifications
Boiling Point 154°C to 155°C
Quantity 5 g
UN Number UN1993
Beilstein 1735027
Formula Weight 160.33
Percent Purity 97%
Refractive Index 1.414
Sensitivity Moisture sensitive
Density 0.816
Chemical Name or Material Ethoxytriethylsilane