missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLP-1R Antibody (7H7), Novus Biologicals™
Human Monoclonal Antibody
€ 355.00 - € 568.00
Specifications
| Antigen | GLP-1R |
|---|---|
| Clone | 7H7 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043101
|
Novus Biologicals
NBP3-26551-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043286
|
Novus Biologicals
NBP3-26551-100ul |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLP-1R Monoclonal antibody specifically detects GLP-1R in Human samples. It is validated for ELISASpecifications
| GLP-1R | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| Recombinant Human GLP-1R protein [UniProt P43220] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7H7 | |
| ELISA | |
| Unconjugated | |
| Human | |
| Cancer, GPCR, Lipid and Metabolism, Signal Transduction | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 2740 | |
| IgG4 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title