missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Secondary Antibody (MR12) (BSA Free), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50261-0.025ml
This item is not returnable.
View return policy
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Immunohistochemistry
Specifications
| IgG | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:10 - 1:500 | |
| Immunoglobulin G, ImmunoglobulinG | |
| Immunoglobulin purified from rabbit serum | |
| 0.025 mL | |
| Secondary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| MR12 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3500 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction