Learn More
Ansell Edmont™ PX140 Series Nylon Light Duty Industrial Gloves
Product Code. p-8394020
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12950005 8 12 pairs
12940005 7 12 pairs
12970005 10 12 pairs
12930005 6 12 pairs
12960005 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12950005 Supplier Ansell Edmont™ Supplier No. C381080
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Comfort and protection for lightweight appications.
  • Industrial gloves
  • Lightweight and stretchable liner for optimized fit and feel
  • Good abrasion resistance due to the polyurethane coating on the palm
  • Elasticated knitwrist to ensure snug fit
  • Comfort and dexterity
  • Ventilated back ensures that the hand can breathe naturally
Applications:
  • Chassis assembly, body trimming
TRUSTED_SUSTAINABILITY
Specifications
Description Not Anti-static, Not silicon-free
Accelerator Free Yes
Applications Industrial
Coating Material Polyurethane
Color White
Disposable Reusable
Exterior Finish No Texture
Latex Free No
Material Polyurethane, Nylon
Powdered Powder Free
Product Type Multi-use Industrial Gloves
Size 8
Sterility Non-sterile
Glove Cuff Style Knit
Liner Material Nylon
Series PX140
Length (English) 8.6 to 10.2 in.
Length (Metric) 220 to 260 mm