missing translation for 'onlineSavingsMsg'
Learn More

Sirtinol, Tocris Bioscience™

Product Code. 18004632
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18004632 10 mg 10mg
18033822 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18004632 Supplier Tocris Supplier No. 3521/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective sirtuin family deacetylase inhibitor

Cell-permeable, selective sirtuin deacetylase inhibitor (IC50 values are 38, 68 and 131 μM at SIRT2, Sir2p and SIRT1 respectively) that has no effect on HDAC1 activity. Significantly decreases growth and viability of PCa and HEK293T cells in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 410536-97-9
Molecular Formula C26H22N2O2
Molecular Weight (g/mol) 394.47
MDL Number MFCD00810186
InChI Key YUGODMKHHCZZOI-ZVTCDHROSA-N
Synonym 2-e-2-hydroxynaphthalen-1-yl methylidene amino-n-1-phenylethyl benzamide
PubChem CID 19577289
IUPAC Name 2-({[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]methyl}amino)-N-[(1S)-1-phenylethyl]benzamide
SMILES C[C@H](NC(=O)C1=CC=CC=C1N\C=C1/C(=O)C=CC2=CC=CC=C12)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 394.47
Percent Purity >98%
Chemical Name or Material Sirtinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.