missing translation for 'onlineSavingsMsg'
Learn More

Sodium arsenite, 0.1N Standardized Solution

Product Code. p-7051559
Change view
Click to view available options
Quantity:
500 mL
4 L
Unit Size:
4 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440401 500 mL 500mL
11304288 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440401 Supplier Thermo Scientific Alfa Aesar Supplier No. 041533.AP
Add to basket
This item is not returnable. View return policy

CAS: 7784-46-5 | AsNaO2 | 129.909 g/mol

Sodium arsenite is used as hide preservative, antiseptic, dyeing and soaps. It is used as a stressor to induce production of heat shock proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-46-5
Molecular Formula AsNaO2
Molecular Weight (g/mol) 129.909
MDL Number MFCD00003472
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
PubChem CID 443495
ChEBI CHEBI:29678
IUPAC Name sodium;oxoarsinite
SMILES [O-][As]=O.[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 0.1N Standardized Solution
Linear Formula NaAsO2
UN Number UN1686
Solubility Information Miscible with water. Slightly miscible with alcohol.
Formula Weight 129.91
Concentration or Composition (by Analyte or Components) 0.1N
Odor Odorless
Chemical Name or Material Sodium arsenite, Standardized Solution
Show More Show Less
compliance-icons
Product Identifier
  • Sodium arsenite, 0.1N Standardized Solution
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium arsenite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.