CAS RN 138729-47-2
CAS RN 138729-47-2
IUPAC Name:
(5S)-6-(5-chloropyridin-2-yl)-7-oxo-5H,6H,7H-pyrrolo[3,4-b]pyrazin-5-yl 4-methylpiperazine-1-carboxylate
Synonyms:
eszopiclone lunesta (+)-zopiclone ( )-zopiclone (S)-zopiclone estorra esopiclone zopiclona [inn-spanish] zopiclone [ban:inn:jan]
Molecular Weight (g/mol):
388.81
Molecular Formula:
C17H17ClN6O3
InChi Key:
GBBSUAFBMRNDJC-INIZCTEOSA-N
SMILES:
CN1CCN(CC1)C(=O)O[C@@H]1N(C(=O)C2=NC=CN=C12)C1=CC=C(Cl)C=N1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Eszopiclone
CAS: 138729-47-2 Molecular Formula: C17H17ClN6O3
2
Eszopiclone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.