CAS RN 25973-55-1
IUPAC Name:
2-(2H-1,2,3-benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
Synonyms:
2-(1,2,3-benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
Molecular Weight (g/mol):
351.49
Molecular Formula:
C22H29N3O
InChi Key:
ZMWRRFHBXARRRT-UHFFFAOYSA-N
SMILES:
CCC(C)(C)C1=CC(N2N=C3C=CC=CC3=N2)=C(O)C(=C1)C(C)(C)CC
1
–
4
of
4
results
1
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality