CAS RN 2642-71-9
IUPAC Name:
O,O-diethyl {[(4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl)methyl]sulfanyl}phosphonothioate
Synonyms:
bionex guthion (ethyl) gusation A ethyl gusathion caswell no. 344 ethyl guthion gusathion athyl-gusathion azinfos-ethyl ethyl homolog of guthion
Molecular Weight (g/mol):
345.37
Molecular Formula:
C12H16N3O3PS2
InChi Key:
RQVGAIADHNPSME-UHFFFAOYSA-N
SMILES:
CCOP(=S)(OCC)SCN1N=NC2=CC=CC=C2C1=O
1
–
10
of
10
results
2
EN 12918 Organophosphorus Mixture 439 10 μg/mL in Acetone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
EN 12918 Organophosphorus Mixture 440 100 μg/mL in Acetone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
4
EPA Method 8141 Organophosphorus Pesticide Mixture 428 200 μg/mL in n-Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality