CAS RN 26575-95-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Alisol B 23-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Alisol B 23-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Alisol B 23-acetate, MedChemExpress
MedChemExpress Alisol B 23-acetate (23-Acetylalismol B), a natural triterpenoid, produces protective effects against EE-induced cholestasis, due to FXR-mediated gene regulation.