CAS RN 29608-49-9
CAS RN 29608-49-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Almitrine Bismesylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Almitrine Bismesylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Almitrine Bismesylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Almitrine mesylate, MedChemExpress
MedChemExpress Almitrine mesylate, a peripheral chemoreceptor agonist, inhibits selectively the Ca2+-dependent K+ channel.