CAS RN 5092-09-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Saikogenin A, MedChemExpress
MedChemExpress Saikogenin A, extracted from a Chinese herbal plant called Tsai-Fu, is a dipeptidyl peptidase-IV (DPP-IV) inhibitor.