Learn More
Poulten & Graf GmbH FORTUNA™ Low Form Graduated Cylinder
Poulten & Graf GmbH FORTUNA™ Low Form Graduated Cylinder measures the volume of a liquid using horizontal marked lines.
€ 3.05 - € 129.00
Specifications
Base Type | Hexagonal |
---|---|
Calibration | To Contain |
Certifications/Compliance | EN ISO 4788 |
Class | B |
Type | Graduated Cylinder |
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
15962150
|
Poulten & Graf GmbH
1 3205104 |
100 mL | 2 mL |
€ 20.40
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15922150
|
Poulten & Graf GmbH
1 3204302 |
25 mL | 1 mL |
€ 3.05
Pack of 2 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
15992150
|
Poulten & Graf GmbH
1 3205904 |
500 mL | 10 mL |
€ 75.80
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15952150
|
Poulten & Graf GmbH
1 3204504 |
50 mL | 2 mL |
€ 18.85
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15912160
|
Poulten & Graf GmbH
1 3206104 |
1000 mL | 20 mL |
€ 129.00
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
15982150
|
Poulten & Graf GmbH
1 3205504 |
250 mL | 5 mL |
€ 39.45
Pack of 2 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10311272
|
Poulten & Graf GmbH
1 3205104 |
100 mL | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10188191
|
Poulten & Graf GmbH
1 3205904 |
500 mL | 10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15932150
|
Poulten & Graf GmbH
1 3204304 |
25 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10209721
|
Poulten & Graf GmbH
1 3204504 |
50 mL | 2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10076101
|
Poulten & Graf GmbH
1 3204304 |
25 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10239621
|
Poulten & Graf GmbH
1 3206104 |
1000 mL | 20 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10259811
|
Poulten & Graf GmbH
1 3203704 |
10 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15912150
|
Poulten & Graf GmbH
1 3203704 |
10 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10448611
|
Poulten & Graf GmbH
1 3204302 |
25 mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10701123
|
Poulten & Graf GmbH
1 3205504 |
250 mL | 5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Conformity approved
- Hexagonal base
- Low form short graduation marks
- With tolerance indications
- Amber graduations are acid and alkali resistant
Specifications
Hexagonal | |
EN ISO 4788 | |
Graduated Cylinder | |
Borosilicate Glass |
To Contain | |
B | |
Round | |
Autoclavable |