Learn More
enQuireBio™ Recombinant Human Serine / arginine-rich splicing factor 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/arginine-rich splicing factor 10, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_001177934.1 |
---|---|
Gene ID (Entrez) | 10772 |
Name | Serine / arginine-rich splicing factor 10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16088960
|
enQuireBio™
QP7486-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008970
|
enQuireBio™
QP7486-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16098960
|
enQuireBio™
QP7486-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018970
|
enQuireBio™
QP7486-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028970
|
enQuireBio™
QP7486-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038970
|
enQuireBio™
QP7486-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001177934.1 | |
Serine / arginine-rich splicing factor 10 Protein | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10772 | |
Research Use Only | |
SRSF10 | |
Recombinant Protein | |
E. coli | |
MSRYLRPPNT SLFVRNVADD TRSEDLRREF GRYGPIVDVY VPLDFYTRRP RGFAYVQFED VRDAEDALHN LDRKWICGRQ IEIQFAQGDR KTPNQMKAKE GRNVYSSSRY DDYDRYRRSR SRSYERRRSR SRSFDYNYRR SYSPRNSRPT GRPRRSRSHS DNDRPNCSWN TQYSSAYYTS RKI | |
Greater than 90% as determined by SDS-PAGE. |